Anti-MDC1 Mouse Monoclonal Antibody (Biotin) [clone: P2B11]

Supplier: Abnova
MAB11471
ABNOMAB11471EA 643 EUR
ABNOMAB11471
Anti-MDC1 Mouse Monoclonal Antibody (Biotin) [clone: P2B11]
Antibodies
Mouse monoclonal antibody raised against recombinant partial Mdc1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MDC1
Clonality: Monoclonal
Clone: P2B11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chimpanzee, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR