Anti-CD6 Mouse Monoclonal Antibody [clone: 3F7B5]

Supplier: Abnova
MAB11334
ABNOMAB11334EA 518 EUR
ABNOMAB11334
Anti-CD6 Mouse Monoclonal Antibody [clone: 3F7B5]
Antibodies
Mouse monoclonal antibody raised against CD6.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 1:100-1:200; Immunohistochemistry: 1:100-1:200; 30 min at RT; Immunoprecipitation: 1-2 ?g/500 ?g protein; Western Blot: 1:250-1:500

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 3F7B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR