Anti-CD1A Mouse Monoclonal Antibody [clone: O10]

Supplier: Abnova
ABNOMAB11333EA 518 EUR
ABNOMAB11333
Anti-CD1A Mouse Monoclonal Antibody [clone: O10]
Antibodies
Mouse monoclonal antibody raised against CD1A.

Recommended Dilutions: ELISA Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 1-2 ?g/ml; Immunohistochemistry: 1-2 ?g/ml for 30 min at RT; Immunoprecipitation: 1-2 ?g/500 ?g protein; Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: O10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR