Anti-PGR Mouse Monoclonal Antibody [clone: PR501]

Supplier: Abnova
MAB11328
ABNOMAB11328EA 518 EUR
ABNOMAB11328
Anti-PGR Mouse Monoclonal Antibody [clone: PR501]
Antibodies
Mouse monoclonal antibody raised against recombinant PGR.

Recommended Dilutions: Western Blot: 0.5-1.0?g/ml for 2 hours at Room Temp)

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR