Anti-UPF1 Mouse Monoclonal Antibody [clone: 4G3]

Supplier: Abnova
H00005976-M01A
ABNOH00005976-M01AEA 394 EUR
ABNOH00005976-M01A
Anti-UPF1 Mouse Monoclonal Antibody [clone: 4G3]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant UPF1.

Type: Primary
Antigen: UPF1
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR