Anti-SLC9A9 Mouse Monoclonal Antibody [clone: 1E10]

Supplier: Abnova
ABNOH00285195-M01EA 394 EUR
ABNOH00285195-M01
Anti-SLC9A9 Mouse Monoclonal Antibody [clone: 1E10]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant SLC9A9.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SLC9A9
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR