Anti-OBSCN Mouse Monoclonal Antibody [clone: 5C20]

Supplier: Abnova
H00084033-M02
ABNOH00084033-M02EA 394 EUR
ABNOH00084033-M02
Anti-OBSCN Mouse Monoclonal Antibody [clone: 5C20]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant OBSCN.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: OBSCN
Clonality: Monoclonal
Clone: 5C20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR