Anti-CENPK Mouse Polyclonal Antibody

Supplier: Abnova
H00064105-B01P
ABNOH00064105-B01PEA 394 EUR
ABNOH00064105-B01P
Anti-CENPK Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human CENPK protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CENPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR