Anti-CDK6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB28592
ABNOPAB28592EA 518 EUR
ABNOPAB28592
Anti-CDK6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CDK6.

Recommended Dilutions: Western Blot: 1:100-1:250; Immunofluorescence: 1-4 ?g/ml; Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: CDK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR