Anti-METTL21B Mouse Polyclonal Antibody

Supplier: Abnova
H00025895-B01P
ABNOH00025895-B01PEA 394 EUR
ABNOH00025895-B01P
Anti-METTL21B Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human FAM119B protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: METTL21B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR