Anti-GRIN1 Mouse Monoclonal Antibody (ATTO 594) [clone: S308-48]
Supplier: Abnova
ABNOMAB11714EA 772 EUR
ABNOMAB11714
Anti-GRIN1 Mouse Monoclonal Antibody (ATTO 594) [clone: S308-48]
Antibodies
Mouse monoclonal antibody raised against partial synthetic protein of Grin1.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: GRIN1
Clonality: Monoclonal
Clone: S308-48
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now