Anti-KCNMB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S18A-3]

Supplier: Abnova
MAB11738
ABNOMAB11738EA 545 EUR
ABNOMAB11738
Anti-KCNMB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S18A-3]
Antibodies
Mouse monoclonal antibody raised against partial synthetic protein of Kcnmb4 .

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: KCNMB4
Clonality: Monoclonal
Clone: S18A-3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR