Anti-AQP4 Rabbit Polyclonal Antibody (ATTO 488)
Supplier: Abnova
PAB28894
ABNOPAB28894EA 772 EUR
ABNOPAB28894
Anti-AQP4 Rabbit Polyclonal Antibody (ATTO 488)
Antibodies
Rabbit polyclonal antibody raised against recombinant Rat Aqp4.
Recommended Dilutions: Immunofluorescence: 1:400; Western Blot: 1:1000-4000
Type:
Antigen: AQP4
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now