Anti-KDEL Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Abnova
PAB28974
ABNOPAB28974EA 772 EUR
ABNOPAB28974
Anti-KDEL Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type:
Antigen: KDEL
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR