Anti-CK10 Mouse Monoclonal Antibody [clone: SPM115]

Supplier: Abnova
ABNOMAB12076EA 518 EUR
ABNOMAB12076
Anti-CK10 Mouse Monoclonal Antibody [clone: SPM115]
Antibodies
Mouse monoclonal antibody raised against Acidic Cytokeratin.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CK10
Clonality: Monoclonal
Clone: SPM115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR