Anti-MSN Mouse Monoclonal Antibody [clone: SPM562]

Supplier: Abnova
MAB12110
ABNOMAB12110EA 518 EUR
ABNOMAB12110
Anti-MSN Mouse Monoclonal Antibody [clone: SPM562]
Antibodies
Mouse monoclonal antibody raised against MSN.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: SPM562
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR