Anti-CNTNAP1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S65-35]

Supplier: Abnova
MAB11906
ABNOMAB11906EA 545 EUR
ABNOMAB11906
Anti-CNTNAP1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S65-35]
Antibodies
Mouse monoclonal antibody raised against partial recombinant Cntnap1.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: CNTNAP1
Clonality: Monoclonal
Clone: S65-35
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR