Anti-IgG Chicken Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Abnova
PAB29100
ABNOPAB29100EA 170 EUR
ABNOPAB29100
Anti-IgG Chicken Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Chicken polyclonal antibody raised against Goat IgG. The antibody is conjugated with FITC.

Recommended Dilutions: Immunohistochemistry: 1:250; Western Blot: 1:1000

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR