Anti-IgG Chicken Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Abnova
PAB29107
ABNOPAB29107EA 262 EUR
ABNOPAB29107
Anti-IgG Chicken Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Chicken polyclonal antibody raised against Mouse IgG. The antibody is conjugated with HRP.

Recommended Dilutions: Western Blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR