Anti-SCN8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S87A-10]

Supplier: Abnova
MAB11990
ABNOMAB11990EA 545 EUR
ABNOMAB11990
Anti-SCN8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S87A-10]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of Scn8a.

Recommended Dilutions: Immunocytochemistry (0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: SCN8A
Clonality: Monoclonal
Clone: S87A-10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR