Anti-CA9 Mouse Monoclonal Antibody [clone: SPM487]

Supplier: Abnova
MAB12025
ABNOMAB12025EA 518 EUR
ABNOMAB12025
Anti-CA9 Mouse Monoclonal Antibody [clone: SPM487]
Antibodies
Mouse monoclonal antibody raised against CA9.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: SPM487
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR