Anti-CGB Mouse Monoclonal Antibody [clone: SPM105]

Supplier: Abnova
MAB12039
ABNOMAB12039EA 518 EUR
ABNOMAB12039
Anti-CGB Mouse Monoclonal Antibody [clone: SPM105]
Antibodies
Mouse monoclonal antibody raised against CGB.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: SPM105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR