Anti-ESR1 Mouse Monoclonal Antibody [clone: SPM567]

Supplier: Abnova
MAB12085
ABNOMAB12085EA 518 EUR
ABNOMAB12085
Anti-ESR1 Mouse Monoclonal Antibody [clone: SPM567]
Antibodies
Mouse monoclonal antibody raised against partial recombinant ESR1.

Recommended Dilutions: Immunohistochemistry: 1-2 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: SPM567
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR