Anti-ABCC4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB28670
ABNOPAB28670EA 518 EUR
ABNOPAB28670
Anti-ABCC4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ABCC4.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: ABCC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR