Anti-MAPK1 Mouse Monoclonal Antibody [clone: 1D1]

Supplier: Abnova
ABNOH00005594-M01AEA 394 EUR
ABNOH00005594-M01A
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 1D1]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant MAPK1.

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR