Anti-PICK1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S20-8]

Supplier: Abnova
MAB11635
ABNOMAB11635EA 545 EUR
ABNOMAB11635
Anti-PICK1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S20-8]
Antibodies
Mouse monoclonal antibody raised against partial synthetic protein of Pick1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: PICK1
Clonality: Monoclonal
Clone: S20-8
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR