Anti-GRIA2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S21-32]

Supplier: Abnova
MAB11647
ABNOMAB11647EA 545 EUR
ABNOMAB11647
Anti-GRIA2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S21-32]
Antibodies
Mouse monoclonal antibody raised against partial synthetic protein of Gria2.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: GRIA2
Clonality: Monoclonal
Clone: S21-32
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR