Anti-MAP3K8 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB29208EA 419 EUR
ABNOPAB29208
Anti-MAP3K8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAP3K8.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: MAP3K8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR