Anti-TMEM179B Mouse Monoclonal Antibody [clone: 3G8]
Supplier: Abnova
ABNOH00374395-M01EA 394 EUR
ABNOH00374395-M01
Anti-TMEM179B Mouse Monoclonal Antibody [clone: 3G8]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant TMEM179B.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: TMEM179B
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now