VWR®, Salt activated nuclease
Supplier: VWR Chemicals
1B1664-25KUEA 289 EUR
1B1664-25KU
VWR®, Salt activated nuclease
Enzymes
Salt Active Nuclease (SAN) is a very active, non specific endonuclease from Pichia pastoris that cleaves double- and single-stranded DNA and RNA. It is active at above neutral pH (optimal pH 8,5) over a wide temperature range (10 to 400 °C), and unlike other nucleases it has optimum activity at high concentrations of salt (0,5 M NaCl). SAN is ideal for use in removal of DNA and RNA from cell extracts and protein samples. Thus, it is very useful in recombinant protein purifications.