Anti-Ghrelin Chicken Polyclonal Antibody

Supplier: Biosensis
C-1525-200
BSENC-1525-200EA 383 EUR
BSENC-1525-200
Anti-Ghrelin Chicken Polyclonal Antibody
Antibodies
Ghrelin is the ligand for growth hormone secretagogue receptor type 1 (GHSR) and upon binding to the receptor it induces the release of growth hormone from the pituitary. This ligand has an appetite-stimulating effect and is involved in growth regulation (Ref: SWISSPROT).

The antibody reacts with human Ghrelin and has been specifically tested in ELISA against the peptide. The epitope used to make this antibody maps to the active portion of Ghrelin 27 or Ghrelin 28, amino acids (24-33), thus this protein will detect active Ghrelin in non-extracted solutions.
Application Information:
ELISA . Suggested dilution at 1:500 to 1:4,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Ghrelin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Maps to amino acids 24-33 of Human Ghrelin
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR