Anti-Arg3.1 Rabbit Polyclonal Antibody
Supplier: Biosensis
BSENR-154-100EA 302 EUR
BSENR-154-100
Anti-Arg3.1 Rabbit Polyclonal Antibody
Antibodies
Arc (also termed activity-regulated cytoskeleton-associated protein or Arg3.1), is an effector immediate early gene whose upregulation has been demonstrated during events of synaptic plasticity. Arg3.1 expression is detectable in neuronal cell bodies and dendrites in the brain regions including striatum and cortex hippocampus, hypothalamus, amygdala.
Antibody is known to recognize Arg3.1 and has been tested in IHC. A dilution of 1:50 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Other applications and other species have not been tested.
Application Information:
IHC. A dilution of 1: 50 to 1: 1000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Arg3.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C terminal of Human Arg3.1 protein
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse