Certificats
Structure chimique
Données du fabricant
Connexion
/
S'inscrire
Belgium
Products
Services
Digital Solutions
Applications
Support
About
0,00 €
Order Entry
Accueil
Tiges de statif
Retort rods, M 10
Retort rods, M 10
Retort rods, M 10
:
R L ENTERPRISES
Retort rods, M 10
:
R L ENTERPRISES
Stainless steel.
Seq
Ø
Longueur
Cdt
241-0188
3108/13
01
10 mm
500 mm
1
1 * 1 ST
1 * 1 ST - 57,30 €
57,30 €
-
+
1 * 1 ST
57,30 €
-
+
Specifications
Environmentally Preferable
RLAA3108/33
3108/33
02
12,5 mm
750 mm
1
1 * 1 ST
1 * 1 ST - 70,90 €
70,90 €
-
+
1 * 1 ST
70,90 €
-
+
Specifications
Environmentally Preferable
RLAA3108/43
3108/43
03
12,5 mm
1000 mm
1
1 * 1 ST
1 * 1 ST - 78,90 €
78,90 €
-
+
1 * 1 ST
78,90 €
-
+
Specifications
Environmentally Preferable