DNA ladder, 100 bp Plus, DNAmark™
Supplier: G-Biosciences
GENO786-856EA 182 EUR
GENO786-856
DNA ladder, 100 bp Plus, DNAmark™
Electrophoresis Reagents Molecular Weight Markers
DNAmark™ 100 bp Plus ladder consists of 14 DNA fragments ranging in size from 100 to 3000 base pairs (bp). The 500 bp and 1200 bp fragment are present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 500 bp and 1200 bp are 40 ng. The 500 bp and 1200 bp fragment are 100 ng.
  • Pre-mixed with loading dye and ready to use