Anti-MAP3K5 Antibody Pair

Supplier: Abnova
DP0039
ABNODP0039EA 735 EUR
ABNODP0039
Anti-MAP3K5 Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the MAP3K5 protein, and the other against the specific S83 phosphorylated site of MAP3K5 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-MAP3K5 S83 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. MAP3K5 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: MAP3K5
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR