Anti-ATM Antibody Pair

Supplier: Abnova
DP0076
ABNODP0076EA 735 EUR
ABNODP0076
Anti-ATM Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the ATM protein, and the other against the specific S1981 phosphorylated site of ATM for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-ATM S1981 Rabbit Polyclonal Antibody (20 µl) In PBS (0.09% (w/v) sodium azide) 2. ATM Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: ATM
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR