Anti-RGS5 Antibody Pair

Supplier: Abnova
ABNOH00008490-AP11EA 835 EUR
ABNOH00008490-AP11
Anti-RGS5 Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human RGS5.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-RGS5 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-RGS5, IgG2b kappa (20 µg);

Type:
Antigen: RGS5
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR