Anti-RPS6KB1 Antibody Pair

Supplier: Abnova
ABNODP0225EA 735 EUR
ABNODP0225
Anti-RPS6KB1 Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the RPS6KB1 protein, and the other against the specific S424 phosphorylated site of RPS6KB1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-RPS6KB1 S424 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. RPS6KB1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: RPS6KB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR