Anti-AKT1 Antibody Pair

Supplier: Abnova
DP0246
ABNODP0246EA 735 EUR
ABNODP0246
Anti-AKT1 Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the AKT1 protein, and the other against the specific T450 phosphorylated site of AKT1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-AKT1 T450 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. AKT1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: AKT1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR