Anti-KLK3 + A2M Antibody Pair

Supplier: Abnova
ABNODI0036EA 668 EUR
ABNODI0036
Anti-KLK3 + A2M Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the KLK3 protein, and the other against the A2M protein for use in Proximity Ligation Assay

Antibody pair set content: 1. KLK3 Rabbit purified Polyclonal Antibody (20 µg) 2. A2M Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: KLK3 + A2M
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR