Anti-TXNDC3 Polyclonal Antibody Pair
Supplier: Abnova
ABNOH00051314-AP21EA 835 EUR
ABNOH00051314-AP21
Anti-TXNDC3 Polyclonal Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human TXNDC3.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-TXNDC3 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-TXNDC3 (20 µg);
Type:
Antigen: TXNDC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human