Anti-RET + MAPK1 Antibody Pair

Supplier: Abnova
DI0281
ABNODI0281EA 668 EUR
ABNODI0281
Anti-RET + MAPK1 Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the RET protein, and the other against the MAPK1 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. RET Rabbit purified Polyclonal Antibody (20 µg) 2. MAPK1 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: RET + MAPK1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR