Anti-TP53 + Laminin, Alpha 4 Polyclonal Antibody Pair

Supplier: Abnova
DI0561
ABNODI0561EA 668 EUR
ABNODI0561
Anti-TP53 + Laminin, Alpha 4 Polyclonal Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the TP53 protein, and the other against the LAMA4 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. TP53 Rabbit purified Polyclonal Antibody (20 µg) 2. LAMA4 Mouse purified Polyclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: TP53 + Laminin, Alpha 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR