Anti-EIF4E Polyclonal Antibody Pair

Supplier: Abnova
DP0268
ABNODP0268EA 735 EUR
ABNODP0268
Anti-EIF4E Polyclonal Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the EIF4E protein, and the other against the specific S209 phosphorylated site of EIF4E for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-EIF4E S209 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. EIF4E Mouse purified Polyclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR