Anti-TAGLN Mouse Monoclonal Antibody (Biotin)

Supplier: Abnova
H00006876-AP44
ABNOH00006876-AP44EA 835 EUR
ABNOH00006876-AP44
Anti-TAGLN Mouse Monoclonal Antibody (Biotin)
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human TAGLN.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-TAGLN (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-TAGLN, IgG2a kappa (50 ?g);

Type: Tag
Antigen: TAGLN
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR