Anti-GRN Mouse Monoclonal Antibody (Biotin)

Supplier: Abnova
H00002896-AP41
ABNOH00002896-AP41EA 835 EUR
ABNOH00002896-AP41
Anti-GRN Mouse Monoclonal Antibody (Biotin)
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human GRN.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-GRN (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-GRN, IgG2a kappa (50 ?g);

Type:
Antigen: GRN
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR