Imprimer
Colorimeters, single test, 1200 series
Fournisseur: LaMotte
LAMO3627-510EA 681 EUR
LAMO3627-510 LAMO3627-530 LAMO3627-570 LAMO3627-460
Colorimeters, single test, 1200 series
Colorimètres Colorimètres monoparamètres
1200 series of single wavelength absorbance colorimeter.
  • Water resistant design with large 3 digit display presents measurements in absorbance, and indicates low battery warnings
  • Two sample vial options with six different wavelengths are available
  • Microprocessor enables the factory programmed calibrations to optimally match non-linear curves
  • With RS232 interface and auto-zero option
The flip-top lid over sample chamber prevents any stray light, especially in the field, and avoids misplacing separate light caps.
Information de livraison: Supplied with vials and water sample collecting bottles in a sturdy carrying case.