Digital Solutions
Applications
Support
About
Order Entry
Western blot roller, Pierce™
PIER84747
:
Thermo Fisher Scientific
Western blot roller, Pierce™
PIER84747
:
Thermo Fisher Scientific
58,25 €
1 * 1 ST
-
+
Désignation:
Western blot roller, PTFE, 8,4 mm wide, with stainless steel handle
Environmentally Preferable:
Cdt:
1