Imprimer
Ultra-low temperature chest freezers, HERAfreeze™ HFC series, −40°C
Fournisseur: Thermo Scientific
HERAHFC2050TVEA 0 EUR
HERAHFC2050TV HERAHFC1750TV
Ultra-low temperature chest freezers, HERAfreeze™ HFC series, −40°C
These ultra-low temperature chest freezers are constructed with stainless steel interior and cold-rolled steel exterior with a powder coat paint finish that resists chipping and rust. It provides a full range of racking solutions for maximum sample storage.
  • Access code to ensure setpoint security
  • Easy-to-remove air filter
  • Heavy duty casters with locking tandem wheel
  • Foamed-in-place (127 mm), polyurethane insulation
  • Includes RS232 and 4 to 20 mA analog outputs for independent monitoring devices and recorders