Imprimer
Terbium standard solution, for ICP-MS, Pure Plus grade
Fournisseur: Perkin Elmer
PERCN9303753EA 46.3 EUR
PERCN9303753
Terbium standard solution, for ICP-MS, Pure Plus grade
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Attention: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).